siRNA Details
| Gene Symbol: C5orf66-AS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037895.1 | 4 | 1467 | |||
| GenBank mRNA | BC045187.1 | - | 4 | 1469 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C5orf66-AS2| Gene Symbol: C5orf66-AS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037895.1 | 4 | 1467 | |||
| GenBank mRNA | BC045187.1 | - | 4 | 1469 | ||