siRNA Details
Gene Symbol: LINC01013 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038981.1 | 1 | 494 | |||
GenBank mRNA | AF086187.1 | - | 1 | 61 | ||
BC040870.1 | - | 1 | 494 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC01013 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038981.1 | 1 | 494 | |||
GenBank mRNA | AF086187.1 | - | 1 | 61 | ||
BC040870.1 | - | 1 | 494 |