siRNA Details
| Gene Symbol: AIFM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001018060.2 | NP_001018070.1 | 10 | 1041 | ||
| NM_001146288.1 | NP_001139760.1 | 10 | 1126 | |||
| NM_144704.2 | NP_653305.1 | 10 | 1041 | |||
| NR_027464.1 | 9 | 998 | ||||
| GenBank mRNA | AK055035.1 | - | 10 | 1025 | ||
| AK094844.1 | - | 10 | 1030 | |||
| AK295581.1 | - | 9 | 901 | |||
| AK316042.1 | - | 9 | 894 | |||
| AY927574.1 | - | 1 | 158 | |||
| BC032485.1 | - | 10 | 1008 | 5221203 | ||
| CR456342.1 | - | 9 | 860 | |||

AIFM3
Genomic Map