siRNA Details
| Gene Symbol: YDJC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017964.1 | NP_001017964.1 | 5 | 1265 | ||
| XM_011529906.2 | XP_011528208.1 | 4 | 1362 | |||
| GenBank mRNA | BC096754.1 | - | 5 | 1338 | ||
| BC110799.1 | - | 5 | 1309 | |||
| BQ050386.1 | - | 1 | 234 | |||

YDJC
Genomic Map