siRNA Details
| Gene Symbol: HSCB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318314.1 | NP_001305243.1 | 3 | 438 | ||
| NM_001318315.1 | NP_001305244.1 | 3 | 438 | |||
| NM_001318316.1 | NP_001305245.1 | 3 | 187 | |||
| NM_172002.4 | NP_741999.3 | 3 | 438 | |||
| NR_134560.1 | 3 | 383 | ||||
| XM_005261359.3 | XP_005261416.1 | 3 | 439 | |||
| XM_017028619.1 | XP_016884108.1 | 3 | 404 | |||
| XM_017028620.1 | XP_016884109.1 | 3 | 242 | |||
| GenBank mRNA | AY191719.1 | - | 3 | 388 | ||
| BC000004.1 | - | 3 | 447 | |||
| BC065569.1 | - | 3 | 411 | 6171959 | ||
| BG696148.1 | - | 3 | 372 | |||
| BQ054405.1 | - | 3 | 415 | |||
| CR456462.1 | - | 3 | 406 | |||
| CR979598.1 | - | 3 | 387 | |||
| DW414316.1 | - | 3 | 203 | |||
| HY057929.1 | - | 3 | 187 | |||

HSCB
Genomic Map