siRNA Details
| Gene Symbol: HORMAD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011529917.2 | XP_011528219.1 | 10 | 1129 | ||
| GenBank mRNA | AK098703.1 | - | 10 | 1095 | ||
| BC030013.1 | - | 10 | 880 | 4823215 | ||
| CR456341.1 | - | 9 | 857 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HORMAD2| Gene Symbol: HORMAD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011529917.2 | XP_011528219.1 | 10 | 1129 | ||
| GenBank mRNA | AK098703.1 | - | 10 | 1095 | ||
| BC030013.1 | - | 10 | 880 | 4823215 | ||
| CR456341.1 | - | 9 | 857 | |||