siRNA Details
| Gene Symbol: NFAM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318323.1 | NP_001305252.1 | 2 | 240 | ||
| NM_145912.6 | NP_666017.1 | 2 | 240 | |||
| XM_005261374.4 | XP_005261431.1 | 4 | 867 | |||
| GenBank mRNA | AY121370.1 | - | 2 | 240 | ||
| AY247409.1 | - | 2 | 235 | |||
| BC038241.1 | - | 2 | 235 | 5180790 | ||
| BI906855.1 | - | 2 | 214 | |||
| CT841509.1 | - | 2 | 240 | |||

NFAM1
Genomic Map