siRNA Details
| Gene Symbol: NFAM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318323.1 | NP_001305252.1 | 5 | 695 | ||
| NM_145912.6 | NP_666017.1 | 6 | 808 | |||
| XM_005261374.4 | XP_005261431.1 | 8 | 1435 | |||
| GenBank mRNA | AK074094.1 | - | 5 | 400 | ||
| AY121370.1 | - | 6 | 808 | |||
| AY247409.1 | - | 6 | 803 | |||
| BC038241.1 | - | 6 | 803 | 5180790 | ||
| CT841509.1 | - | 6 | 808 | |||

NFAM1
Genomic Map