siRNA Details
| Gene Symbol: TEKT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286559.1 | NP_001273488.1 | 1 | 625 | ||
| NM_144705.3 | NP_653306.1 | 1 | 625 | |||
| XM_005263876.2 | XP_005263933.1 | 1 | 4291 | |||
| XM_011510670.1 | XP_011508972.1 | 1 | 4291 | |||
| GenBank mRNA | AK097438.1 | - | 1 | 625 | ||
| AK296678.1 | - | 1 | 550 | |||
| BC021716.2 | - | 1 | 571 | 4831157 | ||
| BI832036.1 | - | 1 | 616 | |||

TEKT4
Genomic Map