siRNA Details
Gene Symbol: LINC01104 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103730.1 | 1 | 541 | |||
GenBank mRNA | AK056640.1 | - | 1 | 541 | ||
BC105017.1 | - | 1 | 145 | |||
BC105019.1 | - | 1 | 145 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC01104 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103730.1 | 1 | 541 | |||
GenBank mRNA | AK056640.1 | - | 1 | 541 | ||
BC105017.1 | - | 1 | 145 | |||
BC105019.1 | - | 1 | 145 |