siRNA Details
| Gene Symbol: OR6B3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173351.1 | NP_775486.1 | 1 | 766 | ||
| XM_011510671.2 | XP_011508973.1 | 3 | 1163 | |||
| Gene Symbol: OR6B2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005853.1 | NP_001005853.1 | 1 | 766 | ||
| GenBank mRNA | BC137561.1 | - | 1 | 826 | 9021561 | |
| BC137562.1 | - | 1 | 826 | 9021562 | ||

OR6B3
Genomic Map