siRNA Details
| Gene Symbol: ITPRIPL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008949.2 | NP_001008949.1 | 3 | 699 | ||
| NM_001163523.1 | NP_001156995.1 | 2 | 589 | |||
| NM_001163524.1 | NP_001156996.1 | 2 | 456 | |||
| NM_001324490.1 | NP_001311419.1 | 2 | 381 | |||
| NM_178495.5 | NP_848590.3 | 1 | 799 | |||
| XM_017003427.1 | XP_016858916.1 | 2 | 632 | |||
| GenBank mRNA | BC034503.2 | - | 1 | 774 | 4824775 | |
| BC073153.1 | - | 3 | 615 | 6018749 | ||
| CD300165.1 | - | 2 | 376 | |||

ITPRIPL1
Genomic Map