siRNA Details
| Gene Symbol: KLHL23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144711.5 | NP_653312.2 | 2 | 1182 | ||
| GenBank mRNA | AK095131.1 | - | 758 | |||
| BC010437.1 | - | 1016 | 3354344 | |||
| Gene Symbol: PHOSPHO2-KLHL23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199290.1 | NP_001186219.1 | 4 | 1313 | ||
| GenBank mRNA | AK090653.1 | - | 4 | 1276 | ||
| AX746559.1 | - | 4 | 1276 | |||

KLHL23
Genomic Map