siRNA Details
| Gene Symbol: SULT1C2P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037191.1 | 2 | 249 | |||
| GenBank mRNA | AK056906.1 | - | 2 | 249 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SULT1C2P1| Gene Symbol: SULT1C2P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037191.1 | 2 | 249 | |||
| GenBank mRNA | AK056906.1 | - | 2 | 249 | ||