siRNA Details
| Gene Symbol: SLC23A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001144889.1 | NP_001138361.1 | 3 | 459 | ||
| NM_001144890.1 | NP_001138362.1 | 3 | 459 | |||
| NM_144712.4 | NP_653313.3 | 3 | 459 | |||
| GenBank mRNA | AK055730.1 | - | 3 | 328 | ||
| AK298256.1 | - | 3 | 369 | |||
| AK298277.1 | - | 3 | 368 | |||
| BC030243.1 | - | 3 | 357 | 5188653 | ||
| DC361649.1 | - | 3 | 462 | |||
| DQ294736.1 | - | 1 | 33 | |||

SLC23A3
Genomic Map