siRNA Details
| Gene Symbol: FAM19A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005527.2 | NP_001005527.1 | 3 | 490 | ||
| NM_182522.4 | NP_872328.1 | 3 | 572 | |||
| XM_011533371.1 | XP_011531673.1 | 3 | 572 | |||
| XM_011533372.1 | XP_011531674.1 | 3 | 448 | |||
| GenBank mRNA | AK057890.1 | - | 2 | 188 | ||
| AY325117.1 | - | 3 | 544 | |||
| BC031566.1 | - | 3 | 404 | 5195201 | ||

FAM19A4
Genomic Map