siRNA Details
| Gene Symbol: EFHB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144715.3 | NP_653316.3 | 4 | 1285 | ||
| XM_005264889.1 | XP_005264946.1 | 4 | 1285 | |||
| XM_011533382.1 | XP_011531684.1 | 3 | 1222 | |||
| XM_011533383.2 | XP_011531685.2 | 4 | 393 | |||
| XM_011533385.2 | XP_011531687.1 | 5 | 661 | |||
| XM_017005741.1 | XP_016861230.1 | 6 | 845 | |||
| XM_017005742.1 | XP_016861231.1 | 4 | 1285 | |||
| XM_017005743.1 | XP_016861232.1 | 5 | 612 | |||
| XM_017005744.1 | XP_016861233.1 | 4 | 524 | |||
| XM_017005745.1 | XP_016861234.1 | 5 | 587 | |||
| GenBank mRNA | AK097644.1 | - | 4 | 1287 | ||
| AK122616.1 | - | 6 | 821 | |||
| BC028198.1 | - | 4 | 1251 | 5209454 | ||

EFHB
Genomic Map