siRNA Details
Gene Symbol: DPPA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138815.3 | NP_620170.3 | 6 | 688 | ||
XM_011512443.1 | XP_011510745.1 | 6 | 529 | |||
XM_011512444.2 | XP_011510746.1 | 6 | 671 | |||
XM_011512445.2 | XP_011510747.1 | 6 | 554 | |||
GenBank mRNA | AY283672.1 | - | 6 | 690 | ||
BC018070.1 | - | 6 | 566 | 4817054 |
