siRNA Details
| Gene Symbol: XXYLT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001308069.1 | NP_001294998.1 | 2, 3 | 306 | ||
| NM_152531.4 | NP_689744.3 | 2, 3 | 769 | |||
| XM_005269286.4 | XP_005269343.1 | 2, 3 | 3154 | |||
| XM_011512448.2 | XP_011510750.1 | 3, 4 | 581 | |||
| XM_017005749.1 | XP_016861238.1 | 2, 3 | 762 | |||
| GenBank mRNA | AK075551.1 | - | 2, 3 | 737 | ||
| AK301133.1 | - | 2, 3 | 242 | |||
| BC019036.2 | - | 2, 3 | 678 | 4591786 | ||
| BC039067.1 | - | 2, 3 | 751 | 5185557 | ||
| BG721419.1 | - | 2, 3 | 311 | |||

XXYLT1
Genomic Map