siRNA Details
| Gene Symbol: RNF38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022781.4 | NP_073618.3 | 3 | 456 | ||
| NM_194328.2 | NP_919309.1 | 3 | 623 | |||
| NM_194329.2 | NP_919310.1 | 2 | 306 | |||
| NM_194330.2 | NP_919311.1 | 3 | 354 | |||
| NM_194332.2 | NP_919313.1 | 2 | 473 | |||
| XM_005251366.3 | XP_005251423.1 | 3 | 460 | |||
| XM_005251367.3 | XP_005251424.1 | 3 | 358 | |||
| XM_005251368.3 | XP_005251425.1 | 3 | 472 | |||
| XM_006716721.3 | XP_006716784.1 | 2 | 320 | |||
| XM_011517712.2 | XP_011516014.1 | 4 | 396 | |||
| XM_011517713.2 | XP_011516015.1 | 3 | 460 | |||
| XM_017014295.1 | XP_016869784.1 | 3 | 487 | |||
| GenBank mRNA | AF394047.1 | - | 3 | 578 | ||
| AK097653.1 | - | 3 | 322 | |||
| AL136817.1 | - | 3 | 374 | |||
| BC033786.1 | - | 2 | 306 | 5166325 | ||
| BG722531.1 | - | 2 | 478 | |||
| BI520161.1 | - | 3 | 352 | |||
| DA189510.1 | - | 2 | 424 | |||
| DA483270.1 | - | 2 | 204 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_175201, NM_001038993, BC062976, BC060730, AK044324, and others.

RNF38
Genomic Map