siRNA Details
| Gene Symbol: EPHA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017000534.1 | XP_016856023.1 | 6 | 1678 | ||
| XM_017000535.1 | XP_016856024.1 | 6 | 1638 | |||
| XM_017000536.1 | XP_016856025.1 | 5 | 1486 | |||
| XM_017000537.1 | XP_016856026.1 | 6 | 1580 | |||
| GenBank mRNA | AK296788.1 | - | 5 | 731 | ||
| BC037166.2 | - | 6 | 1526 | 5090654 | ||
| EU826606.1 | - | 6 | 1408 | |||
| M59371.1 | - | 6 | 1521 | |||

EPHA2
Genomic Map