siRNA Details
| Gene Symbol: EPHA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017000534.1 | XP_016856023.1 | 3 | 556 | ||
| XM_017000535.1 | XP_016856024.1 | 3 | 516 | |||
| XM_017000536.1 | XP_016856025.1 | 2 | 364 | |||
| XM_017000537.1 | XP_016856026.1 | 3 | 458 | |||
| GenBank mRNA | BC037166.2 | - | 3 | 404 | 5090654 | |
| CB991812.1 | - | 3 | 468 | |||
| EU826606.1 | - | 3 | 286 | |||
| HY005615.1 | - | 3 | 464 | |||

EPHA2
Genomic Map