siRNA Details
Gene Symbol: BRCA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007294.3 | NP_009225.1 | 12 | 4430 | ||
NM_007297.3 | NP_009228.2 | 11 | 4338 | |||
NM_007298.3 | NP_009229.2 | 11 | 908 | |||
NM_007299.3 | NP_009230.2 | 12 | 1083 | |||
NM_007300.3 | NP_009231.2 | 12 | 4430 | |||
NR_027676.1 | 12 | 4334 | ||||
GenBank mRNA | AF005068.1 | - | 12 | 4299 | ||
AK316200.1 | - | 11 | 980 | |||
AY751490.1 | - | 11 | 4198 | |||
BC046142.1 | - | 6 | 514 | |||
BC072418.1 | - | 12 | 1083 | |||
BC085615.1 | - | 12 | 988 | |||
BC115037.1 | - | 12 | 4335 | 7961446 | ||
JN686490.1 | - | 11 | 4198 | |||
U14680.1 | - | 12 | 4317 | |||
U64805.1 | - | 11 | 889 |

More Information
Matching Taqman Gene Expression Assay:
Panther Classification:
Gene Ontology Categories:
