siRNA Details
| Gene Symbol: NFXL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278623.1 | NP_001265552.1 | 20 | 2424 | ||
| NM_001278624.1 | NP_001265553.1 | 20 | 2562 | |||
| NM_152995.5 | NP_694540.3 | 20 | 2464 | |||
| NR_103795.1 | 21 | 2604 | ||||
| GenBank mRNA | AB085695.1 | - | 20 | 2420 | ||
| AB181916.1 | - | 20 | 2544 | |||
| AK131285.1 | - | 18 | 2010 | |||
| AK302440.1 | - | 20 | 2422 | |||
| AY134856.1 | - | 4 | 468 | |||
| BC018019.1 | - | 3 | 159 | |||
| BC051193.1 | - | 21 | 2575 | 5271401 | ||

NFXL1
Genomic Map