siRNA Details
| Gene Symbol: THAP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317791.1 | NP_001304720.1 | 4 | 544 | ||
| NM_144721.5 | NP_653322.1 | 5 | 670 | |||
| XM_005262772.2 | XP_005262829.1 | 4 | 565 | |||
| XM_011531666.2 | XP_011529968.1 | 5 | 1107 | |||
| XM_011531667.2 | XP_011529969.1 | 5 | 630 | |||
| XM_017007800.1 | XP_016863289.1 | 4 | 710 | |||
| GenBank mRNA | AK313765.1 | - | 5 | 1054 | ||
| AL833350.1 | - | 5 | 630 | |||
| BC022989.2 | - | 4 | 584 | 5113206 | ||
| BX647579.1 | - | 4 | 504 | |||

THAP6
Genomic Map