siRNA Details
| Gene Symbol: PPM1K | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152542.4 | NP_689755.3 | 4 | 1162 | ||
| XM_006714111.3 | XP_006714174.1 | 4 | 1223 | |||
| XM_017007803.1 | XP_016863292.1 | 4 | 810 | |||
| XM_017007804.1 | XP_016863293.1 | 5 | 1350 | |||
| XM_017007805.1 | XP_016863294.1 | 6 | 1350 | |||
| XM_017007806.1 | XP_016863295.1 | 4 | 683 | |||
| XM_017007807.1 | XP_016863296.1 | 3 | 401 | |||
| GenBank mRNA | AK054678.1 | - | 4 | 890 | ||
| AK314417.1 | - | 4 | 887 | |||
| AL834271.1 | - | 4 | 881 | |||
| AX766363.1 | - | 3 | 677 | |||
| AY157615.1 | - | 4 | 890 | |||
| AY435431.1 | - | 4 | 1067 | |||
| AY994097.1 | - | 4 | 898 | |||
| BC037552.1 | - | 4 | 926 | 5227317 | ||

PPM1K
Genomic Map