siRNA Details
| Gene Symbol: CEP120 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166226.1 | NP_001159698.1 | 5 | 627 | ||
| NM_153223.3 | NP_694955.2 | 6 | 708 | |||
| XM_005271901.4 | XP_005271958.1 | 5 | 624 | |||
| XM_011543185.2 | XP_011541487.1 | 5 | 1042 | |||
| XM_011543186.2 | XP_011541488.1 | 2 | 193 | |||
| XM_017009085.1 | XP_016864574.1 | 2 | 193 | |||
| GenBank mRNA | AK095646.1 | - | 5 | 629 | ||
| AK296999.1 | - | 6 | 672 | |||
| BC040527.1 | - | 5 | 938 | |||
| BC144558.1 | - | 8 | 835 | |||
| CR627324.1 | - | 1 | 1075 | |||

CEP120
Genomic Map