siRNA Details
| Gene Symbol: C19orf71 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135580.1 | NP_001129052.1 | 4 | 586 | ||
| XM_011527596.2 | XP_011525898.1 | 4 | 512 | |||
| GenBank mRNA | BX396051.2 | - | 4 | 589 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C19orf71| Gene Symbol: C19orf71 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135580.1 | NP_001129052.1 | 4 | 586 | ||
| XM_011527596.2 | XP_011525898.1 | 4 | 512 | |||
| GenBank mRNA | BX396051.2 | - | 4 | 589 | ||