siRNA Details
| Gene Symbol: FAM47E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136570.2 | NP_001130042.1 | 8 | 1515 | ||
| NM_001242936.1 | NP_001229865.1 | 8 | 1588 | |||
| GenBank mRNA | AK124936.1 | - | 8 | 1588 | ||
| CR627383.1 | - | 6 | 6191 | |||

FAM47E
Genomic Map