siRNA Details
| Gene Symbol: CCDC152 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134848.1 | NP_001128320.1 | 9 | 3268 | ||
| GenBank mRNA | BC039102.1 | - | 9 | 3240 | ||
| DB515085.1 | - | 1 | 211 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CCDC152| Gene Symbol: CCDC152 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134848.1 | NP_001128320.1 | 9 | 3268 | ||
| GenBank mRNA | BC039102.1 | - | 9 | 3240 | ||
| DB515085.1 | - | 1 | 211 | |||