siRNA Details
| Gene Symbol: LRRC70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181506.4 | NP_852607.3 | 2 | 376 | ||
| GenBank mRNA | AK123022.1 | - | 2 | 376 | ||
| AY280614.1 | - | 2 | 221 | |||
| AY358733.1 | - | 2 | 240 | |||
| BC110838.1 | - | 2 | 228 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LRRC70| Gene Symbol: LRRC70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181506.4 | NP_852607.3 | 2 | 376 | ||
| GenBank mRNA | AK123022.1 | - | 2 | 376 | ||
| AY280614.1 | - | 2 | 221 | |||
| AY358733.1 | - | 2 | 240 | |||
| BC110838.1 | - | 2 | 228 | |||