siRNA Details
| Gene Symbol: LRRC70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181506.4 | NP_852607.3 | 2 | 667 | ||
| GenBank mRNA | AK123022.1 | - | 2 | 667 | ||
| AY280614.1 | - | 2 | 512 | |||
| AY358733.1 | - | 2 | 531 | |||
| BC067275.1 | - | 1 | 196 | |||
| BC110838.1 | - | 2 | 519 | |||

LRRC70
Genomic Map