siRNA Details
| Gene Symbol: PATE3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001129883.3 | NP_001123355.3 | 3 | 688 | ||
| XM_017017054.1 | XP_016872543.1 | 2 | 3988 | |||
| GenBank mRNA | EF426753.1 | - | 3 | 688 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PATE3| Gene Symbol: PATE3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001129883.3 | NP_001123355.3 | 3 | 688 | ||
| XM_017017054.1 | XP_016872543.1 | 2 | 3988 | |||
| GenBank mRNA | EF426753.1 | - | 3 | 688 | ||