siRNA Details
| Gene Symbol: PLEKHG4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052909.3 | NP_443141.3 | 5 | 921 | ||
| XM_006714446.3 | XP_006714509.1 | 7 | 2059 | |||
| XM_011513968.2 | XP_011512270.1 | 7 | 2059 | |||
| XM_011513969.2 | XP_011512271.1 | 7 | 1936 | |||
| XM_011513970.1 | XP_011512272.1 | 7 | 1648 | |||
| XM_011513971.2 | XP_011512273.1 | 7 | 2004 | |||
| XM_011513972.2 | XP_011512274.1 | 7 | 2060 | |||
| XM_011513973.1 | XP_011512275.1 | 7 | 2057 | |||
| XM_017009112.1 | XP_016864601.1 | 7 | 2060 | |||
| GenBank mRNA | AB067496.2 | - | 5 | 921 | ||
| BC008352.1 | - | 5 | 613 | |||

PLEKHG4B
Genomic Map