siRNA Details
| Gene Symbol: ZMAT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144723.2 | NP_653324.1 | 5 | 523 | ||
| GenBank mRNA | AK055683.1 | - | 5 | 436 | ||
| BC056668.1 | - | 5 | 440 | 4773709 | ||
| BP210985.1 | - | 5 | 517 | |||
| BP370587.1 | - | 4 | 379 | |||

ZMAT2
Genomic Map