siRNA Details
| Gene Symbol: BTNL9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152547.4 | NP_689760.2 | 11 | 2671 | ||
| XM_006714823.3 | XP_006714886.1 | 10 | 2646 | |||
| XM_006714824.3 | XP_006714887.1 | 10 | 2646 | |||
| XM_006714825.3 | XP_006714888.1 | 12 | 3058 | |||
| XM_017009113.1 | XP_016864602.1 | 9 | 2625 | |||
| XM_017009114.1 | XP_016864603.1 | 11 | 2538 | |||
| XM_017009116.1 | XP_016864605.1 | 6 | 2228 | |||
| XM_017009117.1 | XP_016864606.1 | 5 | 2207 | |||
| GenBank mRNA | AL833663.1 | - | 1 | 1175 | ||
| AY358358.1 | - | 11 | 2656 | |||
| BC029040.1 | - | 6 | 1573 | |||
| BC042112.1 | - | 6 | 1573 | |||

BTNL9
Genomic Map