siRNA Details
| Gene Symbol: ARSK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198150.2 | NP_937793.1 | 8 | 1561 | ||
| XM_005271904.3 | XP_005271961.1 | 9 | 1625 | |||
| GenBank mRNA | AK303855.1 | - | 8 | 1613 | ||
| AL832711.1 | - | 7 | 1244 | |||
| AY358596.1 | - | 8 | 1521 | |||
| AY875939.1 | - | 8 | 1403 | |||
| BC036047.1 | - | 8 | 1423 | |||
| BC050392.1 | - | 9 | 1571 | |||
| BC130329.1 | - | 8 | 1499 | 40146360 | ||
| BC144161.1 | - | 9 | 1558 | |||

ARSK
Genomic Map