siRNA Details
| Gene Symbol: PTPMT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001143984.1 | NP_001137456.1 | 2 | 694 | ||
| NM_175732.2 | NP_783859.1 | 4 | 777 | |||
| GenBank mRNA | AF277187.1 | - | 2 | 191 | ||
| AY333987.1 | - | 3 | 392 | |||
| BC014048.1 | - | 3 | 423 | |||
| BC020242.1 | - | 4 | 606 | 4591438 | ||
| BC107573.1 | - | 2 | 529 | |||
| BQ050653.1 | - | 2 | 170 | |||
| DR004030.1 | - | 4 | 635 | |||

PTPMT1
Genomic Map