siRNA Details
| Gene Symbol: FAM81B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152548.2 | NP_689761.2 | 5 | 646 | ||
| XM_005271906.4 | XP_005271963.2 | 6 | 669 | |||
| XM_005271907.3 | XP_005271964.1 | 5 | 631 | |||
| XM_011543207.1 | XP_011541509.1 | 5 | 641 | |||
| XM_011543208.1 | XP_011541510.1 | 5 | 641 | |||
| XM_011543209.1 | XP_011541511.1 | 3 | 327 | |||
| XM_017009118.1 | XP_016864607.1 | 6 | 666 | |||
| GenBank mRNA | AK058062.1 | - | 5 | 629 | ||
| AK310245.1 | - | 4 | 448 | |||
| BC034772.1 | - | 5 | 646 | 5169904 | ||
| BM560335.1 | - | 4 | 451 | |||

FAM81B
Genomic Map