siRNA Details
| Gene Symbol: IFFO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136265.1 | NP_001129737.1 | 9 | 1901 | ||
| XM_011540630.2 | XP_011538932.1 | 8 | 2100 | |||
| GenBank mRNA | AK024480.1 | - | 5 | 923 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

IFFO2| Gene Symbol: IFFO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136265.1 | NP_001129737.1 | 9 | 1901 | ||
| XM_011540630.2 | XP_011538932.1 | 8 | 2100 | |||
| GenBank mRNA | AK024480.1 | - | 5 | 923 | ||