siRNA Details
| Gene Symbol: VWA3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173615.4 | NP_775886.3 | 16 | 1559 | ||
| XM_011545742.2 | XP_011544044.1 | 19 | 1769 | |||
| XM_011545744.2 | XP_011544046.1 | 4 | 369 | |||
| XM_011545745.1 | XP_011544047.1 | 2 | 137 | |||
| XM_017022947.1 | XP_016878436.1 | 20 | 1842 | |||
| XM_017022948.1 | XP_016878437.1 | 3 | 941 | |||
| XM_017022949.1 | XP_016878438.1 | 2 | 907 | |||
| GenBank mRNA | AK098260.1 | - | 14 | 1516 | ||
| AK128606.1 | - | 5 | 427 | |||
| AK307544.1 | - | 17 | 1618 | |||
| BC109294.1 | - | 4 | 287 | |||
| BC109295.1 | - | 4 | 287 | |||

VWA3A
Genomic Map