siRNA Details
| Gene Symbol: VWA3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173615.4 | NP_775886.3 | 3 | 276 | ||
| XM_011545742.2 | XP_011544044.1 | 4 | 369 | |||
| XM_017022947.1 | XP_016878436.1 | 5 | 442 | |||
| GenBank mRNA | AK098260.1 | - | 3 | 273 | ||
| AK307544.1 | - | 3 | 272 | |||

VWA3A
Genomic Map