siRNA Details
| Gene Symbol: SMIM13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135575.1 | NP_001129047.1 | 2 | 4519 | ||
| GenBank mRNA | AJ420516.1 | - | 1 | 1653 | ||
| AK129879.1 | - | 1 | 1472 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SMIM13| Gene Symbol: SMIM13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135575.1 | NP_001129047.1 | 2 | 4519 | ||
| GenBank mRNA | AJ420516.1 | - | 1 | 1653 | ||
| AK129879.1 | - | 1 | 1472 | |||