siRNA Details
| Gene Symbol: ZSCAN23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012455.1 | NP_001012458.1 | 4 | 1498 | ||
| GenBank mRNA | AL833347.1 | - | 4 | 1463 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZSCAN23| Gene Symbol: ZSCAN23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012455.1 | NP_001012458.1 | 4 | 1498 | ||
| GenBank mRNA | AL833347.1 | - | 4 | 1463 | ||