siRNA Details
Gene Symbol: KIAA1107 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017000800.1 | XP_016856289.1 | 4 | 935 | ||
GenBank mRNA | AB029030.1 | - | 3 | 706 | ||
AK302496.1 | - | 3 | 811 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: KIAA1107 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017000800.1 | XP_016856289.1 | 4 | 935 | ||
GenBank mRNA | AB029030.1 | - | 3 | 706 | ||
AK302496.1 | - | 3 | 811 |