siRNA Details
| Gene Symbol: SYCE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105578.1 | NP_001099048.1 | 4 | 335 | ||
| XM_005259848.4 | XP_005259905.1 | 4 | 407 | |||
| XM_011527882.2 | XP_011526184.1 | 4 | 737 | |||
| XM_011527883.2 | XP_011526185.1 | 4 | 737 | |||
| GenBank mRNA | AK097443.1 | - | 3 | 1588 | ||
| AK302373.1 | - | 4 | 325 | |||
| BC035819.2 | - | 3 | 304 | |||
| DB449708.1 | - | 4 | 340 | |||

SYCE2
Genomic Map