siRNA Details
| Gene Symbol: C22orf24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001302819.1 | NP_001289748.1 | 2 | 670 | ||
| NM_001302820.1 | NP_001289749.1 | 2 | 597 | |||
| NM_015372.2 | NP_056187.1 | 3 | 461 | |||
| NR_126507.1 | 3 | 388 | ||||
| GenBank mRNA | AJ704765.1 | - | 3 | 412 | ||
| AJ704766.1 | - | 3 | 339 | |||
| AK294232.1 | - | 2 | 640 | |||
| AK315264.1 | - | 3 | 459 | |||
| AL050256.1 | - | 3 | 449 | |||
| DB102820.1 | - | 3 | 388 | |||

C22orf24
Genomic Map