siRNA Details
| Gene Symbol: C22orf24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001302819.1 | NP_001289748.1 | 2 | 861 | ||
| NM_001302820.1 | NP_001289749.1 | 2 | 788 | |||
| NM_015372.2 | NP_056187.1 | 3 | 652 | |||
| NR_126507.1 | 3 | 579 | ||||
| GenBank mRNA | AJ704765.1 | - | 3 | 603 | ||
| AJ704766.1 | - | 3 | 530 | |||
| AK294232.1 | - | 2 | 831 | |||
| AK315264.1 | - | 3 | 650 | |||
| AL050256.1 | - | 3 | 640 | |||

C22orf24
Genomic Map