siRNA Details
| Gene Symbol: PRELID2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138492.5 | NP_612501.3 | 5 | 541 | ||
| NM_182960.3 | NP_892005.1 | 5 | 374 | |||
| NM_205846.2 | NP_995318.1 | 4 | 338 | |||
| XM_017009127.1 | XP_016864616.1 | 5 | 374 | |||
| XM_017009128.1 | XP_016864617.1 | 5 | 374 | |||
| XM_017009129.1 | XP_016864618.1 | 5 | 374 | |||
| XM_017009130.1 | XP_016864619.1 | 5 | 374 | |||
| XM_017009131.1 | XP_016864620.1 | 5 | 374 | |||
| XM_017009132.1 | XP_016864621.1 | 5 | 374 | |||
| XM_017009133.1 | XP_016864622.1 | 5 | 374 | |||
| XM_017009134.1 | XP_016864623.1 | 5 | 374 | |||
| XM_017009135.1 | XP_016864624.1 | 4 | 338 | |||
| GenBank mRNA | AK095695.1 | - | 5 | 309 | ||
| AK125215.1 | - | 4 | 273 | |||
| BC012033.1 | - | 5 | 508 | 4476359 | ||
| BC028175.1 | - | 255 | ||||
| BQ645893.1 | - | 4 | 308 | |||
| BU167299.1 | - | 4 | 200 | |||
| BU186346.1 | - | 5 | 311 | |||

PRELID2
Genomic Map