siRNA Details
| Gene Symbol: C20orf194 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009984.2 | NP_001009984.1 | 16 | 1436 | ||
| XM_005260684.3 | XP_005260741.1 | 16 | 1430 | |||
| XM_017027783.1 | XP_016883272.1 | 13 | 1057 | |||
| GenBank mRNA | AK226100.1 | - | 11 | 860 | ||
| AK303375.1 | - | 7 | 805 | |||
| AL110249.1 | - | 7 | 401 | |||
| BC122529.1 | - | 7 | 654 | 40012183 | ||

C20orf194
Genomic Map